Learn More
Biolegend PE/Cyanine7 anti-human CD360 I
Catalog No. 502722138
Encompass_Preferred
PE/Cyanine7 anti-human CD360 (IL-21R) [17A12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2138 Supplier Biolegend Supplier No. 359514
PE/Cyanine7 anti-human CD360 (IL-21R) [17A12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests