Learn More
Biolegend PE/Cyanine7 anti-human CD365 (Tim-1)
Supplier:  Biolegend 353910
Encompass_Preferred
PE/Cyanine7 anti-human CD365 (Tim-1) [1D12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-7297