Learn More
Biolegend PE/Cyanine7 anti-human CD45
Supplier:  Biolegend 368531
Encompass_Preferred
PE/Cyanine7 anti-human CD45 [2D1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8431