Learn More
Biolegend PE/Cyanine7 anti-human CD5
Supplier:  Biolegend 364008
Encompass_Preferred
PE/Cyanine7 anti-human CD5 [L17F12]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8195