Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Human CD56 (NCAM) (MY31) ..... Cat. No. 60-0564-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 600564T025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Human CD56 (NCAM) (MY31) ..... Cat. No. 60-0564-T025 ..... Size 25 testsThe MY31 antibody reacts with human CD56, also known as the neural cell adhesion molecule (NCAM), a glycoprotein which is a member of the immunoglobulin superfamily. The 140 kDa isoform of CD56 is expressed on human NK cells and NK-T cells, with increased expression levels on activated NK lymphocytes. The CD56 antigen is also expressed by neurons and is reported to play a role in nervous system development and neural cell-to-cell adhesion. Clone MY31 also reacts with a subset of CD14+ monocytes in non-human primates, and is reported to be cross-reactive with Chimpanzee, Cynomolgus and Rhesus.
Catalog No. 50-210-3527