Learn More
Biolegend PE/Cyanine7 anti-human CD64
Catalog No. 502721567
Encompass_Preferred
PE/Cyanine7 anti-human CD64 [10.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 μL
Catalog No. 50-272-1567 Supplier Biolegend Supplier No. 983208
PE/Cyanine7 anti-human CD64 [10.1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 μL