Learn More
Biolegend PE/Cyanine7 anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Supplier:  Biolegend 985106
Encompass_Preferred
PE/Cyanine7 anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-237-5069