Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Human HLA-DR (LN3) ..... Cat. No. 60-9956-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 609956T025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Human HLA-DR (LN3) ..... Cat. No. 60-9956-T025 ..... Size 25 testsThe LN3 antibody reacts with a member of the human MHC Class II antigens, HLA-DR. The HLA-DR antigen is expressed on human antigen presenting cells including B lymphocytes, monocytes, macrophages, dendritic cells, and activated T lymphocytes. HLA-DR is a dimeric protein composed of alpha and beta subunits and is involved in the presentation of peptide antigens to CD4+ T cells.
Catalog No. 50-210-3552