Learn More
Biolegend PE/Cyanine7 anti-human TCR Vde
Catalog No. 502722300
Encompass_Preferred
PE/Cyanine7 anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2300 Supplier Biolegend Supplier No. 331421
PE/Cyanine7 anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests