Learn More
Biolegend PE/Cyanine7 anti-mouse CD191 C
Catalog No. 502722612
Encompass_Preferred
PE/Cyanine7 anti-mouse CD191 (CCR1) [S15040E]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2612 Supplier Biolegend Supplier No. 152509
PE/Cyanine7 anti-mouse CD191 (CCR1) [S15040E]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg