Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Mouse CD45.2 (104) ..... Cat. No. 60-0454-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 600454U025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Mouse CD45.2 (104) ..... Cat. No. 60-0454-U025 ..... Size 25 µgThe 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.
Catalog No. 50-210-3523