Learn More
Cytek Biosciences Inc PE-Cy7 aMu CD8a Anti Primary
Supplier: Cytek Biosciences Inc 601886U025
Product Name: PE-Cyanine7 Anti-Mouse CD8a (2.43)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Mouse CD8a (2.43)
Cytek Catalog Number: 60-1886-U025
Marker: CD8a
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Monoclonal
Clone: 2.43
Host Species: Rat
Isotype: IgG2b
Conjugation: PE-Cyanine7
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.