Learn More
Biolegend PE/Cyanine7 anti-mouse PLZF
Supplier:  Biolegend 145806
Encompass_Preferred
PE/Cyanine7 anti-mouse PLZF [9E12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-207-1671