Learn More
Biolegend PE/Cyanine7 anti-mouse CD105
Supplier:  Biolegend 120409
Encompass_Preferred
PE/Cyanine7 anti-mouse CD105 [MJ7/18]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0394