Learn More
Biolegend PE/Cyanine7 anti-mouse CD11a
Supplier:  Biolegend 101121
Encompass_Preferred
PE/Cyanine7 anti-mouse CD11a [M17/4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1902237