Learn More
Biolegend PE/Cyanine7 anti-mouse CD133
Supplier:  Biolegend 141210
Encompass_Preferred
PE/Cyanine7 anti-mouse CD133 [315-2C11]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1338