Learn More
Biolegend PE/Cyanine7 anti-mouse CD159a (NKG2AB6)
Supplier:  Biolegend 142809
Encompass_Preferred
PE/Cyanine7 anti-mouse CD159a (NKG2AB6) [16A11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1415