Learn More
Biolegend PE/Cyanine7 anti-mouse CD160
Supplier:  Biolegend 143009
Encompass_Preferred
PE/Cyanine7 anti-mouse CD160 [7H1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1420