Learn More
Biolegend PE/Cyanine7 anti-mouse CD223 (LAG-3)
Supplier:  Biolegend 125226
Encompass_Preferred
PE/Cyanine7 anti-mouse CD223 (LAG-3) [C9B7W]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0656