Learn More
Biolegend PE/Cyanine7 anti-mouse CD43
Supplier:  Biolegend 143209
Encompass_Preferred
PE/Cyanine7 anti-mouse CD43 [S11]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1428