Learn More
Biolegend PE/Cyanine7 anti-mouse IgG2b
Supplier:  Biolegend 406714
Encompass_Preferred
PE/Cyanine7 anti-mouse IgG2b [RMG2b-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-9794