Learn More
Biolegend PE/Cyanine7 anti-mouse Ly-51
Supplier:  Biolegend 108313
Encompass_Preferred
PE/Cyanine7 anti-mouse Ly-51 [6C3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0022