Learn More
Biolegend PE/Cyanine7 anti-mouse Tim-4
Supplier:  Biolegend 130010
Encompass_Preferred
PE/Cyanine7 anti-mouse Tim-4 [RMT4-54]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0901