Learn More
Biolegend PE/Cyanine7 anti-OCT4 OCT3
Catalog No. 503107980
Encompass_Preferred
PE/Cyanine7 anti-OCT4 (OCT3) [3A2A20] Isotype Mouse IgG2b Reactivity Human Mouse Apps ICFC Size 100 tests
Catalog No. 50-310-7980 Supplier Biolegend Supplier No. 653718
PE/Cyanine7 anti-OCT4 (OCT3) [3A2A20] Isotype Mouse IgG2b Reactivity Human Mouse Apps ICFC Size 100 tests