Learn More
Biolegend PE/Cyanine7 anti-OCT4 OCT3
Catalog No. 503108075
Encompass_Preferred
PE/Cyanine7 anti-OCT4 (OCT3) [3A2A20] Isotype Mouse IgG2b Reactivity Human Mouse Apps ICFC Size 25 tests
Catalog No. 50-310-8075 Supplier Biolegend Supplier No. 653717
PE/Cyanine7 anti-OCT4 (OCT3) [3A2A20] Isotype Mouse IgG2b Reactivity Human Mouse Apps ICFC Size 25 tests