Learn More
Biolegend PE/Cyanine7 anti-rat IgM
Supplier:  Biolegend 408914
Encompass_Preferred
PE/Cyanine7 anti-rat IgM [MRM-47]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-207-9856