Learn More
Biolegend PE/Dazzle™ 594 anti-human CD199 (CCR9)
Supplier:  Biolegend 358918
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD199 (CCR9) [L053E8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1775780