Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD1d (CD1.1, Ly-38)
Supplier:  Biolegend 123520
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1538004