Learn More
Biolegend PE/Dazzle™ 594 anti-human CD141 (Thrombomodulin)
Supplier:  Biolegend 344120
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD141 (Thrombomodulin) [M80]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests
Catalog No. NC1474065