Learn More
Biolegend PE/Dazzle™ 594 anti-human CD25
Supplier:  Biolegend 356126
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD25 [M-A251]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1275604