Learn More
Biolegend PE/Dazzle™ 594 anti-human CD366 (Tim-3)
Supplier:  Biolegend 345034
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD366 (Tim-3) [F38-2E2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1249754