Learn More
Biolegend PE/Dazzle™ 594 anti-human CD4
Supplier:  Biolegend 357411
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD4 [A161A1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1373881