Learn More
Biolegend PE/Dazzle™ 594 anti-human CD8
Supplier:  Biolegend 344744
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: African Green, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests
Catalog No. NC1776338