Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD152
Supplier:  Biolegend 106317
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1851600