Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD3
Supplier:  Biolegend 100246
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1052008