Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD4
Supplier:  Biolegend 100456
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1268504