Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD62L
Supplier:  Biolegend 104448
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1014119