Learn More
Biolegend PE/Dazzle™ 594 anti-mouse GPR15/BOB [S15042I]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Supplier:  Biolegend 154610
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse GPR15/BOB [S15042I]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-237-8473