Learn More
Biolegend PE/Dazzle™ 594 anti-mouse TIGIT (Vstm3)
Supplier:  Biolegend 142110
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse TIGIT (Vstm3) [1G9]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1032563