Learn More
Biolegend CD170, also known as Siglec-F, Siglec-5
Supplier:  Biolegend 155530
Encompass_Preferred
CD170, also known as Siglec-F, Siglec-5, is a member of the Sialic acid-binding Ig-like lectin family, type I single pass transmembrane protein, with 4 extracellular Ig-like domains and 2 ITIM motifs in the cytoplasmic domain; preferentially binds [alpha]-2,3-linked sialic acid. Siglec F is expressed in eosinophils, alveolar macrophages and intestinal microfold (M) cells and induces apoptosis of the lung eosinophis during allergic asthma. 100ug. BIOLEGEND mfg# 155530
Catalog No. NC2083235