Learn More
Biolegend PE/Dazzle™ 594 anti-T-bet
Catalog No. NC1356388
Encompass_Preferred
PE/Dazzle™ 594 anti-T-bet [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 ug
Catalog No. NC1356388 Supplier Biolegend Supplier No. 644828
PE/Dazzle™ 594 anti-T-bet [4B10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 ug