Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD314 (NKG2D)
Supplier:  Biolegend 130214
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD314 (NKG2D) [CX5]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1775699