Learn More
Biolegend PE/DazzleTM 594 anti-human C
Supplier:  Biolegend 343926
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD166 [3A6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-253-7908