Learn More
Biolegend PE/DazzleTM 594 anti-human CD1
Supplier:  Biolegend 302128
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD18 [TS1/18]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2500