Learn More
Biolegend PE/DazzleTM 594 anti-human CD6
Supplier:  Biolegend 305032
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD64 [10.1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-272-2462