Learn More
Biolegend PE/DazzleTM 594 anti-human CD8
Supplier: Biolegend 301058
PE/Dazzle™ 594 anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests