Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD86 [A17199A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Supplier:  Biolegend 159210
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD86 [A17199A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-272-3418