Learn More
Biolegend PE/Dazzle™ 594 anti-human β2-microglobulin
Supplier:  Biolegend 316319
Encompass_Preferred
PE/Dazzle™ 594 anti-human β2-microglobulin [2M2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 25 tests
Catalog No. 50-207-3730