Learn More
Biolegend PE/Dazzle™ 594 anti-human CD11b
Supplier:  Biolegend 301348
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD11b [ICRF44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests
Catalog No. 50-850-478