Learn More
Biolegend PE/Dazzle™ 594 anti-human CD11c
Supplier:  Biolegend 337228
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD11c [Bu15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-103-0734